missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASEF Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11569-1-AP-20UL
This item is not returnable.
View return policy
Description
Binds predomitly GDP, and also GTP.Specifications
| RASEF | |
| Polyclonal | |
| Unconjugated | |
| RASEF | |
| RAB45, Ras related protein Rab 45, RASEF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 158158, 242505 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5RI75, Q8IZ41 | |
| RASEF | |
| RASEF Fusion Protein Ag2149 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction