missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASA4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | RASA4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605900
|
Novus Biologicals
NBP2-93505-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695300
|
Novus Biologicals
NBP2-93505-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RASA4 Polyclonal antibody specifically detects RASA4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RASA4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| B Cell Development and Differentiation Markers, GPCR, Immunology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 10156 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Ca2+-promoted Ras inactivator, Calcium-promoted Ras inactivator, CAPRIGAPLKIAA0538RASA4B, FLJ59070, Putative Ras GTPase-activating protein 4B, ras GTPase-activating protein 4, RAS p21 protein activator 4MGC131890, RasGAP-activating-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-290 of human RASA4 (NP_008920.5). HLPPTFHAVAFYVMDEDALSRDDVIGKVCLTRDTIASHPKGFSGWAHLTEVDPDEEVQGEIHLRLEVWPGARACRLRCSVLEARDLAPKDRNGTSDPFVRVRYKGRTRETSIVKKSCYPRWNETFEFELQEGAMEALCVEAWDWDLVSRNDFLGKVVIDVQRLRVVQQEEGWFRLQPDQSKSRRHDEGNLGSLQLEVRLRDETVLPSSYYQPLVHLLCHEVKLGMQGPGQL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts