missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASA4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93505-0.02ml
This item is not returnable.
View return policy
Description
RASA4 Polyclonal antibody specifically detects RASA4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RASA4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Ca2+-promoted Ras inactivator, Calcium-promoted Ras inactivator, CAPRIGAPLKIAA0538RASA4B, FLJ59070, Putative Ras GTPase-activating protein 4B, ras GTPase-activating protein 4, RAS p21 protein activator 4MGC131890, RasGAP-activating-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-290 of human RASA4 (NP_008920.5). HLPPTFHAVAFYVMDEDALSRDDVIGKVCLTRDTIASHPKGFSGWAHLTEVDPDEEVQGEIHLRLEVWPGARACRLRCSVLEARDLAPKDRNGTSDPFVRVRYKGRTRETSIVKKSCYPRWNETFEFELQEGAMEALCVEAWDWDLVSRNDFLGKVVIDVQRLRVVQQEEGWFRLQPDQSKSRRHDEGNLGSLQLEVRLRDETVLPSSYYQPLVHLLCHEVKLGMQGPGQL | |
| 0.02 mL | |
| B Cell Development and Differentiation Markers, GPCR, Immunology, Signal Transduction | |
| 10156 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction