Learn More
Invitrogen™ RARS Recombinant Rabbit Monoclonal Antibody (4Y1H4)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | RARS |
|---|---|
| Clone | 4Y1H4 |
| Concentration | 1.48 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284793
|
Invitrogen™
MA557989 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283846
![]() |
Invitrogen™
MA557990 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MDVLVSECSA RLLQQEEEIK SLTAEIDRLK NCGCLGASPN LEQLQEENLK LKYRLNILRK SLQAERNKPT KNMINIISRL QEVFGHAIKA AYPDLENPPL LVTPSQQAKF GDYQCNSAMG ISQMLKTKEQ KVNPREIAEN ITKHLPDNEC IEKVEIAGPG FINVHLRKDF VSEQLTSLLV NGVQLPALGE NKKVIVDFSS PNIAKEMHVG HLRSTIIGES ISRLFEFAGY DVLRLNHVGD WGTQFGMLIA HLQDKFPDYL TVSPPIGDLQ VFYKESKKRF DTEEEFKKRA YQCVVLLQGK.
The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. ArgRS (Arginyl-tRNA synthetase), also known as RARS or DALRD1, belongs to the class-I aminoacyl-tRNA synthetase family that includes the related proteins, LeuRS, ValRS and IleRS. These proteins are large monomeric proteins and play a major role in catalyzing the aminoacylation of tRNA by their cognate amino acid. ArgRS localizes to the cytoplasm and exists as a monomer but can also associate with other tRNA synthetases and auxiliary proteins to form a multisubunit complex. In the presence of ATP, arginine (Arg) and tRNA, ArgRS joins Arg to tRNA(Arg) at its synthetic active site. Two cytoplasmic forms of ArgRS have been described in mammals, differing by the addition of a 73 amino acid sequence that is required for ArgRS assembly into the multisubunit complex.Specifications
| RARS | |
| 1.48 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P54136, Q9D0I9 | |
| 104458, 5917 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RARS (NP_002878.2). | |
| Antibody |
| 4Y1H4 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| RARS1 | |
| 2610011N19Rik; 2610037E21Rik; AL033339; arginine tRNA ligase 1, cytoplasmic; arginine--tRNA ligase, cytoplasmic; Arginyl-tRNA synthetase; arginyl-tRNA synthetase, cytoplasmic; argRS; AW985894; DALRD1; HLD9; MGC8641; RARS | |
| RARS1 | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.