missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RARS Recombinant Rabbit Monoclonal Antibody (4Y1H4)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557990

 View more versions of this product

Product Code. 30283846

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MDVLVSECSA RLLQQEEEIK SLTAEIDRLK NCGCLGASPN LEQLQEENLK LKYRLNILRK SLQAERNKPT KNMINIISRL QEVFGHAIKA AYPDLENPPL LVTPSQQAKF GDYQCNSAMG ISQMLKTKEQ KVNPREIAEN ITKHLPDNEC IEKVEIAGPG FINVHLRKDF VSEQLTSLLV NGVQLPALGE NKKVIVDFSS PNIAKEMHVG HLRSTIIGES ISRLFEFAGY DVLRLNHVGD WGTQFGMLIA HLQDKFPDYL TVSPPIGDLQ VFYKESKKRF DTEEEFKKRA YQCVVLLQGK.

The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. ArgRS (Arginyl-tRNA synthetase), also known as RARS or DALRD1, belongs to the class-I aminoacyl-tRNA synthetase family that includes the related proteins, LeuRS, ValRS and IleRS. These proteins are large monomeric proteins and play a major role in catalyzing the aminoacylation of tRNA by their cognate amino acid. ArgRS localizes to the cytoplasm and exists as a monomer but can also associate with other tRNA synthetases and auxiliary proteins to form a multisubunit complex. In the presence of ATP, arginine (Arg) and tRNA, ArgRS joins Arg to tRNA(Arg) at its synthetic active site. Two cytoplasmic forms of ArgRS have been described in mammals, differing by the addition of a 73 amino acid sequence that is required for ArgRS assembly into the multisubunit complex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RARS
Recombinant Monoclonal
1.48 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P54136, Q9D0I9
RARS1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RARS (NP_002878.2).
100 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
4Y1H4
Unconjugated
RARS1
2610011N19Rik; 2610037E21Rik; AL033339; arginine tRNA ligase 1, cytoplasmic; arginine--tRNA ligase, cytoplasmic; Arginyl-tRNA synthetase; arginyl-tRNA synthetase, cytoplasmic; argRS; AW985894; DALRD1; HLD9; MGC8641; RARS
Rabbit
Protein A
RUO
104458, 5917
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.