missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapsyn Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £375.00
Specifications
| Antigen | Rapsyn |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626392
|
Novus Biologicals
NBP2-94107-0.02ml |
0.02 mL |
£159.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653762
|
Novus Biologicals
NBP2-94107-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rapsyn Polyclonal antibody specifically detects Rapsyn in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Rapsyn | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 5913 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 43 kDa postsynaptic protein, Acetylcholine receptor-associated 43 kDa protein, CMS1D43 kDa receptor-associated protein of the synapse, CMS1EMGC3597, RAPsyn, receptor-associated protein of the synapse, receptor-associated protein of the synapse, 43kD, RNF205RING finger protein 205 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-353 of human RAPSN (NP_116034.2). MGQDQTKQQIEKGLQLYQSNQTEKALQVWTKVLEKSSDLMGRFRVLGCLVTAHSEMGRYKEMLKFAVVQIDTARELEDADFLLESYLNLARSNEKLCEFHKTISYCKTCLGLPGTRAGAQLGGQVSLSMGNAFLGLSVFQKALESFEKALRYAHNNDDAMLECRVCCSLGSFYAQVKDYEKALFFPCKAAELVNNYGKGWSLKYRAMSQYHMAVAYRLLGRLGSAMECCEESMKIALQHGDRPLQALCLLCFADIHRSRGDLELSQLKLHCLSESIYRSKGLQRELRAHVVRFHECVEETELYCGLCGESIGEKNSRLQALPCSHIFHLRCLQNNGTRSCPNCRRSSMKPGFV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title