missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapsyn Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94107-0.02ml
This item is not returnable.
View return policy
Description
Rapsyn Polyclonal antibody specifically detects Rapsyn in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Rapsyn | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| 43 kDa postsynaptic protein, Acetylcholine receptor-associated 43 kDa protein, CMS1D43 kDa receptor-associated protein of the synapse, CMS1EMGC3597, RAPsyn, receptor-associated protein of the synapse, receptor-associated protein of the synapse, 43kD, RNF205RING finger protein 205 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-353 of human RAPSN (NP_116034.2). MGQDQTKQQIEKGLQLYQSNQTEKALQVWTKVLEKSSDLMGRFRVLGCLVTAHSEMGRYKEMLKFAVVQIDTARELEDADFLLESYLNLARSNEKLCEFHKTISYCKTCLGLPGTRAGAQLGGQVSLSMGNAFLGLSVFQKALESFEKALRYAHNNDDAMLECRVCCSLGSFYAQVKDYEKALFFPCKAAELVNNYGKGWSLKYRAMSQYHMAVAYRLLGRLGSAMECCEESMKIALQHGDRPLQALCLLCFADIHRSRGDLELSQLKLHCLSESIYRSKGLQRELRAHVVRFHECVEETELYCGLCGESIGEKNSRLQALPCSHIFHLRCLQNNGTRSCPNCRRSSMKPGFV | |
| 0.02 mL | |
| Neuroscience, Zinc Finger | |
| 5913 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction