missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapsyn Antibody (1234), Novus Biologicals™
Mouse Monoclonal Antibody
£174.00
Specifications
| Antigen | Rapsyn |
|---|---|
| Clone | 1234 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 μg/mL, Immunofluorescence 2 μg/mL |
| Applications | Western Blot, Immunofluorescence |
Description
Rapsyn Monoclonal specifically detects Rapsyn in Human, Mouse, Rat, Amphibian, Chicken, Fish samples. It is validated for Western Blot, Immunofluorescence.Specifications
| Rapsyn | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Amphibian, Chicken, Fish | |
| 5913 | |
| Whole purified rapsyn from Torpedo californica electric organ postsynaptic membrane. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1234 | |
| Western Blot 0.5 μg/mL, Immunofluorescence 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 43 kDa postsynaptic protein, Acetylcholine receptor-associated 43 kDa protein, CMS1D43 kDa receptor-associated protein of the synapse, CMS1EMGC3597, RAPsyn, receptor-associated protein of the synapse, receptor-associated protein of the synapse, 43kD, RNF205RING finger protein 205 | |
| Rapsn | |
| IgG1 | |
| Protein G purified | |
| This antibody reacts with Amphibian, Fish, Mouse, Rat and Chicken. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title