missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapsyn Antibody (1234), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22469-0.025mg
This item is not returnable.
View return policy
Description
Rapsyn Monoclonal specifically detects Rapsyn in Human, Mouse, Rat, Amphibian, Chicken, Fish samples. It is validated for Western Blot, Immunofluorescence.
Specifications
| Rapsyn | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, Immunofluorescence 2 μg/mL | |
| Rapsn | |
| Whole purified rapsyn from Torpedo californica electric organ postsynaptic membrane. | |
| 0.025 mg | |
| Primary | |
| This antibody reacts with Amphibian, Fish, Mouse, Rat and Chicken. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| 1234 | |
| Unconjugated | |
| 43 kDa postsynaptic protein, Acetylcholine receptor-associated 43 kDa protein, CMS1D43 kDa receptor-associated protein of the synapse, CMS1EMGC3597, RAPsyn, receptor-associated protein of the synapse, receptor-associated protein of the synapse, 43kD, RNF205RING finger protein 205 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 5913 | |
| Mouse, Rat, Amphibian, Chicken, Fish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction