missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£222.00 - £451.00
Specifications
| Antigen | RAPH1 |
|---|---|
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18451772
|
Novus Biologicals
NBP1-90859-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18279506
|
Novus Biologicals
NBP1-90859 |
0.1 mL |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAPH1 Polyclonal specifically detects RAPH1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RAPH1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| candidate 18, candidate 9, lamellipodin, PREL2, PREL-2, proline rich EVH1 ligand 2, Protein RMO1, Ras association (RalGDS/AF-6) and pleckstrin homology domains 1, ras-associated and pleckstrin homology domains-containing protein 1, RMO1 | |
| RAPH1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 65059 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NPQPQQWSKMSVKKAPPPTRPKRNDSTRLTQAEISEQPTMATVVPQVPTSPKSSLSVQPGFLADLNRTLQRKSITRHGSLSSRMSRAEPTATMDDMALPPPPPELLSDQQKAGYGGSHISGYATLRRGPPPAPPKRDQNTKLSR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title