missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-90859
This item is not returnable.
View return policy
Description
RAPH1 Polyclonal specifically detects RAPH1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RAPH1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Flow Cytometry, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| candidate 18, candidate 9, lamellipodin, PREL2, PREL-2, proline rich EVH1 ligand 2, Protein RMO1, Ras association (RalGDS/AF-6) and pleckstrin homology domains 1, ras-associated and pleckstrin homology domains-containing protein 1, RMO1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 65059 | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RAPH1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NPQPQQWSKMSVKKAPPPTRPKRNDSTRLTQAEISEQPTMATVVPQVPTSPKSSLSVQPGFLADLNRTLQRKSITRHGSLSSRMSRAEPTATMDDMALPPPPPELLSDQQKAGYGGSHISGYATLRRGPPPAPPKRDQNTKLSR | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction