missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANK/TNFRSF11A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£177.00 - £379.00
Specifications
| Antigen | RANK/TNFRSF11A |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18048323
|
Novus Biologicals
NB100-56396SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18794973
|
Novus Biologicals
NB100-56396 |
0.1 mg |
£379.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RANK/TNFRSF11A Polyclonal specifically detects RANK/TNFRSF11A in Human, Mouse samples. It is validated for Western Blot.Specifications
| RANK/TNFRSF11A | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Q9Y6Q6 | |
| 8792 | |
| A synthetic peptide corresponding to amino acid residues 603-613 of human RANK (NP_003830). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| TNFRSF11A | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title