missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANK/TNFRSF11A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56396SS
This item is not returnable.
View return policy
Description
RANK/TNFRSF11A Polyclonal specifically detects RANK/TNFRSF11A in Human, Mouse samples. It is validated for Western Blot.
Specifications
| RANK/TNFRSF11A | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| Q9Y6Q6 | |
| TNFRSF11A | |
| A synthetic peptide corresponding to amino acid residues 603-613 of human RANK (NP_003830). | |
| 0.025 mg | |
| Cell Cycle and Replication | |
| 8792 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction