missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANBP9 Recombinant Rabbit Monoclonal Antibody (23GB4120), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | RANBP9 |
|---|---|
| Clone | 23GB4120 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089317
|
Invitrogen™
MA553694 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089004
|
Invitrogen™
MA553695 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RANBP9 Recombinant Monoclonal antibody specifically detects RANBP9 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RANBP9 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P69566, Q96S59 | |
| 10048, 364686, 56705 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4120 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B cell antigen receptor Ig beta associated protein 1; B-cell antigen receptor Ig beta-associated protein 1; BPM 90; BPM L; BPM90; BPML; BPM-L; IBAP 1; Ibap1; IBAP-1; Imp 9; Importin 9; novel centrosomal protein RanBPM; RAN binding protein 9; Ran Binding Protein in the Microtubule organizing center; ran binding protein, centrosomal; Ran BP9; ran-binding protein 9; Ran-binding protein M; RanBP 7; RANBP 9; RanBP7; Ranbp9; RANBPM; RGD1561940 | |
| A synthesized peptide derived from human RanBP9 (110-185AA). | |
| Primary | |
| -20°C | |
| RANBP9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title