missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANBP9 Recombinant Rabbit Monoclonal Antibody (23GB4120), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553695
This item is not returnable.
View return policy
Description
This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11.
Specifications
| RANBP9 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RANBP9 | |
| B cell antigen receptor Ig beta associated protein 1; B-cell antigen receptor Ig beta-associated protein 1; BPM 90; BPM L; BPM90; BPML; BPM-L; IBAP 1; Ibap1; IBAP-1; Imp 9; Importin 9; novel centrosomal protein RanBPM; RAN binding protein 9; Ran Binding Protein in the Microtubule organizing center; ran binding protein, centrosomal; Ran BP9; ran-binding protein 9; Ran-binding protein M; RanBP 7; RANBP 9; RanBP7; Ranbp9; RANBPM; RGD1561940 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10048, 364686, 56705 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4120 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P69566, Q96S59 | |
| RANBP9 | |
| A synthesized peptide derived from human RanBP9 (110-185AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction