missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RanBP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00
Specifications
| Antigen | RanBP3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 10 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18577040
|
Novus Biologicals
NB120-2939-50ug |
50 μg | |||||||
Description
RanBP3 Polyclonal specifically detects RanBP3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RanBP3 | |
| Western Blot 10 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 8498 | |
| Synthetic peptide corresponding to residues M(1) A D L A N E E K P A L A(13) C of human RanBP3. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp586I1520, RAN binding protein 3, ran-binding protein 3, RAN-binding protein-3, RanBP3 | |
| RANBP3 | |
| IgG | |
| Affinity Purified | |
| Detects Ran binding protein 3 (RanBP 3) from human and rat cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title