missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RanBP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-2939-50ug
This item is not returnable.
View return policy
Description
RanBP3 Polyclonal specifically detects RanBP3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RanBP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RANBP3 | |
| Synthetic peptide corresponding to residues M(1) A D L A N E E K P A L A(13) C of human RanBP3. | |
| 50 μg | |
| Signal Transduction | |
| 8498 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 10 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| DKFZp586I1520, RAN binding protein 3, ran-binding protein 3, RAN-binding protein-3, RanBP3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects Ran binding protein 3 (RanBP 3) from human and rat cells. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction