missing translation for 'onlineSavingsMsg'
Learn More

RanBP2 Antibody (2E1), Novus Biologicals™

Mouse Monoclonal Antibody

£159.00

Specifications

Antigen RanBP2
Clone 20
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:1000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18563460
View Documents
Novus Biologicals
NB100-74480-0.025mg
0.025 mg
£159.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RanBP2 Monoclonal specifically detects RanBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

RanBP2
1 mg/mL
Western Blot
Unconjugated
Mouse
Human
ANE1, E3 SUMO-protein ligase RanBP2, EC 5.2.1.8, Nuclear pore complex protein Nup358, nucleoporin 358, Nucleoporin Nup358, NUP358TRP2, P270, RAN binding protein 2, Ran-binding protein 2, RanBP2, transformation-related protein 2,358 kDa nucleoporin, TRP1
RANBP2
IgG2b
Protein G purified
Nup358 (2E1)
20
Western Blot 1:100 - 1:1000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000
Monoclonal
Purified
RUO
P49792
5903
Recombinant protein corresponding to 150 N-terminal residues of human Nup358.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.