missing translation for 'onlineSavingsMsg'
Learn More

RanBP2 Antibody (2E1), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-74480-0.025mg

 View more versions of this product

Product Code. 18563460

  • £159.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RanBP2 Monoclonal specifically detects RanBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RanBP2
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:1000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000
ANE1, E3 SUMO-protein ligase RanBP2, EC 5.2.1.8, Nuclear pore complex protein Nup358, nucleoporin 358, Nucleoporin Nup358, NUP358TRP2, P270, RAN binding protein 2, Ran-binding protein 2, RanBP2, transformation-related protein 2,358 kDa nucleoporin, TRP1
Mouse
Protein G purified
RUO
5903
Human
Purified
Western Blot
20
Unconjugated
P49792
RANBP2
Recombinant protein corresponding to 150 N-terminal residues of human Nup358.
0.025 mg
Primary
Nup358 (2E1)
Store at -20°C. Avoid freeze/thaw cycles.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only