missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £309.00
Specifications
| Antigen | RANBP17 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18123215
|
Novus Biologicals
NBP2-23638SS |
0.025 mg |
£155.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18150522
|
Novus Biologicals
NBP2-23638 |
0.1 mg |
£309.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RANBP17 Polyclonal specifically detects RANBP17 in Human samples. It is validated for Western Blot.Specifications
| RANBP17 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 64901 | |
| RanBP17 protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ32916, RAN binding protein 17 | |
| RANBP17 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title