missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23638
This item is not returnable.
View return policy
Description
RANBP17 Polyclonal specifically detects RANBP17 in Human samples. It is validated for Western Blot.
Specifications
| RANBP17 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| FLJ32916, RAN binding protein 17 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 64901 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| RANBP17 | |
| RanBP17 protein | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction