missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ran Antibody (8D1A6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£173.00 - £435.00
Specifications
| Antigen | Ran |
|---|---|
| Clone | 8D1A6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249453
|
Novus Biologicals
NBP2-61832 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662238
|
Novus Biologicals
NBP2-61832-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ran Monoclonal specifically detects Ran in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Ran | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| Androgen receptor-associated protein 24, ARA24Gsp1, GTPase Ran, GTP-binding nuclear protein Ran, guanosine triphosphatase Ran, member RAS oncogene family, OK/SW-cl.81, RAN, member RAS oncogene family, RanGTPase, Ras-like protein TC4, Ras-related nuclear protein, TC4 | |
| RAN | |
| IgG1 | |
| Protein G purified |
| 8D1A6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Monkey | |
| 5901 | |
| Purified recombinant fragment of human RAN (AA: 1-216) expressed in E. Coli. | |
| Primary | |
| 24.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title