missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ran Antibody (8D1A6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-61832-0.025ml
This item is not returnable.
View return policy
Description
Ran Monoclonal specifically detects Ran in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Ran | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Androgen receptor-associated protein 24, ARA24Gsp1, GTPase Ran, GTP-binding nuclear protein Ran, guanosine triphosphatase Ran, member RAS oncogene family, OK/SW-cl.81, RAN, member RAS oncogene family, RanGTPase, Ras-like protein TC4, Ras-related nuclear protein, TC4 | |
| Mouse | |
| 24.4 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 8D1A6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RAN | |
| Purified recombinant fragment of human RAN (AA: 1-216) expressed in E. Coli. | |
| Protein G purified | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 5901 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction