missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Radiation Hazard Trays

Product Code. 12317247 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
54 x 34 cm
70 x 46 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
12317247 54 x 34 cm Each
12347247 70 x 46 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12317247
Only null left
Add to basket
Add to basket

Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.

  • Radiation hazard tray yellow 34 x 54cm
  • Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25

Specifications

Dimensions (L x W) 54 x 34 cm
Color Yellow
Length (Metric) 54 cm
Material APET
Type Radiation Hazard Tray
Width (Metric) 34 cm
For Use With Lab bench
Shape Rectangular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.