missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD50 Recombinant Rabbit Monoclonal Antibody (23GB4115), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | RAD50 |
|---|---|
| Clone | 23GB4115 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089017
![]() |
Invitrogen™
MA553692 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088046
|
Invitrogen™
MA553693 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAD50 Recombinant Monoclonal antibody specifically detects RAD50 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RAD50 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P70388, Q92878, Q9JIL8 | |
| 10111, 19360, 64012 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4115 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 153 kDa protein; DNA repair protein RAD50; hRad50; hsRAD50; I79_004319; mRad50; Mrell; MRX complex DNA-binding subunit; N0872; NBSLD; Rad50; RAD50 2; RAD50 double strand break repair protein; RAD50 homolog (S. cerevisiae); RAD50 homolog, double strand break repair protein; RAD50 PEN; RAD502; RAD50-2; RAD50-3; Rad50l; YNL250W | |
| A synthesized peptide derived from Rad50 (1250-1312AA). | |
| Primary | |
| -20°C | |
| RAD50 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title