missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD50 Recombinant Rabbit Monoclonal Antibody (23GB4115), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553693
This item is not returnable.
View return policy
Description
Rad50-13B3 recognizes human Rad50, a 153a protein involved in DNA double strand break repair. Rad50 is associated with Mre11 and p95 (Nibrin) to form a multiprotein complex involved in the double strand break repair process. Upon DNA damage, Rad50 and Mre11 redistribute and are co-localized within the nucleus of damaged cells.
Specifications
| RAD50 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RAD50 | |
| 153 kDa protein; DNA repair protein RAD50; hRad50; hsRAD50; I79_004319; mRad50; Mrell; MRX complex DNA-binding subunit; N0872; NBSLD; Rad50; RAD50 2; RAD50 double strand break repair protein; RAD50 homolog (S. cerevisiae); RAD50 homolog, double strand break repair protein; RAD50 PEN; RAD502; RAD50-2; RAD50-3; Rad50l; YNL250W | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10111, 19360, 64012 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4115 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70388, Q92878, Q9JIL8 | |
| RAD50 | |
| A synthesized peptide derived from Rad50 (1250-1312AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction