missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD50 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RAD50 |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17402383
|
Proteintech
29390-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17492373
|
Proteintech
29390-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RAD50 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAD50. This antibody recognizes human, rat, mouse antigen. The RAD50 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| RAD50 | |
| Polyclonal | |
| Rabbit | |
| 10111 | |
| Primary |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title