Learn More
RAD50 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29390-1-AP-150UL
Description
The RAD50 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAD50. This antibody recognizes human, rat, mouse antigen. The RAD50 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| RAD50 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| 150 μL | |
| 10111 | |
| IgG |