missing translation for 'onlineSavingsMsg'
Learn More

rac-Lipoic Acid Monosulfoxide (Mixture of Regioisomers and Diastereomers), TRC

Product Code. 30135417
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135417 100 mg 100mg
30133563 1 g 1g
30003325 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135417 Supplier Toronto Research Chemicals Supplier No. TRCL468740100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108015-78-7
Molecular Formula C8 H14 O3 S2
Molecular Weight (g/mol) 222.32
Synonym 1,2-Dithiolane-3-pentanoic acid, mixture of 1-Oxide and 2-Oxide
IUPAC Name 5-(2-oxodithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H14O3S2/c9-8(10)4-2-1-3-7-5-6-12-13(7)11/h7H,1-6H2,(H,9,10)
Formula Weight 222.0384
Chemical Name or Material 5-(2-oxodithiolan-3-yl)pentanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.