missing translation for 'onlineSavingsMsg'
Learn More

rac Ketorolac-d4 (>90%), TRC

Product Code. 30404494
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404494 10 mg 10mg
30401321 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404494 Supplier Toronto Research Chemicals Supplier No. TRCK23560210MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216451-53-4
Molecular Formula C15H9D4NO3
Molecular Weight (g/mol) 259.29
Synonym 5-Benzoyl-2,3-dihydro-1H-(pyrrolizine-d4)-1-carboxylic Acid, (+/-)-Ketorolac-d4, Ketorolac-d4, RS 37619-d4
SMILES O=C(C1=CC=CC=C1)C2=CC=C3N2C([2H])([2H])C([2H])([2H])C3C(O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H13NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13/h1-7,11H,8-9H2,(H,18,19)/i8D2,9D2
Chemical Name or Material rac Ketorolac-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.