CAS RN 1216451-53-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Ketorolac-d4 (>90%), TRC
CAS: 1216451-53-4 Molecular Formula: C15H9D4NO3 Molecular Weight (g/mol): 259.29 Synonym: 5-Benzoyl-2,3-dihydro-1H-(pyrrolizine-d4)-1-carboxylic Acid,(+/-)-Ketorolac-d4,Ketorolac-d4,RS 37619-d4 SMILES: O=C(C1=CC=CC=C1)C2=CC=C3N2C([2H])([2H])C([2H])([2H])C3C(O)=O