missing translation for 'onlineSavingsMsg'
Learn More

rac Ketoprofen-d3, TRC

Product Code. 30397154
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397154 1 mg
30405731 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30397154 Supplier Toronto Research Chemicals Supplier No. TRCK2008031MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159490-55-8
Molecular Formula C16 2H3 H11 O3
Molecular Weight (g/mol) 257.3
Synonym (±)-Ketoprofen D3 (methyl D3), rac Ketoprofen-d3
IUPAC Name 2-(3-benzoylphenyl)-3,3,3-trideuteriopropanoic acid
SMILES [2H]C([2H])([2H])C(C(=O)O)c1cccc(c1)C(=O)c2ccccc2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/i1D3
Formula Weight 257.1131
Percent Purity >95%
Chemical Name or Material (±)-KetoprofenD3(propionicD3acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.