CAS RN 159490-55-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Ketoprofen-d3, TRC
CAS: 159490-55-8 Molecular Formula: C16 2H3 H11 O3 Molecular Weight (g/mol): 257.3 Synonym: (±)-Ketoprofen D3 (methyl D3),rac Ketoprofen-d3 IUPAC Name: 2-(3-benzoylphenyl)-3,3,3-trideuteriopropanoic acid SMILES: [2H]C([2H])([2H])C(C(=O)O)c1cccc(c1)C(=O)c2ccccc2