missing translation for 'onlineSavingsMsg'
Learn More

rac-Hesperetin Triacetate, TRC

Product Code. 30395816
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395816 5 mg
30390138 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395816 Supplier Toronto Research Chemicals Supplier No. TRCH2895205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73489-97-1
Molecular Formula C22H20O9
Molecular Weight (g/mol) 428.39
Synonym (S)-5,7-Bis(acetyloxy)-2-[3-(acetyloxy)-4-methoxyphenyl]-2,3-dihydro-4H-1-benzopyran-4-one, Hesperetin Triacetate, Tri-O-acetylhesperetin
IUPAC Name [(2S)-5-acetyloxy-2-(3-acetyloxy-4-methoxyphenyl)-4-oxo-2,3-dihydrochromen-7-yl] acetate
SMILES COc1ccc(cc1OC(=O)C)[C@@H]2CC(=O)c3c(OC(=O)C)cc(OC(=O)C)cc3O2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H20O9/c1-11(23)28-15-8-20(30-13(3)25)22-16(26)10-18(31-21(22)9-15)14-5-6-17(27-4)19(7-14)29-12(2)24/h5-9,18H,10H2,1-4H3/t18-/m0/s1
Formula Weight 428.1107
Chemical Name or Material rac-Hesperetin Triacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.