CAS RN 73489-97-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac-Hesperetin Triacetate, TRC
CAS: 73489-97-1 Molecular Formula: C22H20O9 Molecular Weight (g/mol): 428.39 Synonym: (S)-5,7-Bis(acetyloxy)-2-[3-(acetyloxy)-4-methoxyphenyl]-2,3-dihydro-4H-1-benzopyran-4-one,Hesperetin Triacetate,Tri-O-acetylhesperetin IUPAC Name: [(2S)-5-acetyloxy-2-(3-acetyloxy-4-methoxyphenyl)-4-oxo-2,3-dihydrochromen-7-yl] acetate SMILES: COc1ccc(cc1OC(=O)C)[C@@H]2CC(=O)c3c(OC(=O)C)cc(OC(=O)C)cc3O2