missing translation for 'onlineSavingsMsg'
Learn More

rac all-trans 4-Hydroxy Retinoic Acid, TRC

Product Code. 30402117
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402117 1 mg -
30408736 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402117 Supplier Toronto Research Chemicals Supplier No. TRCH9533001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66592-72-1
Molecular Formula C20 H28 O3
Molecular Weight (g/mol) 316.43
Synonym Retinoic acid, 4-hydroxy-, 4-Hydroxyretinoic acid, 4-Hydroxy-all-trans-retinoic acid, 4-OH-atRA, all-trans-4-Hydroxyretinoic acid
IUPAC Name (2E,4E,6E,8E)-9-(3-hydroxy-2,6,6-trimethylcyclohexen-1-yl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
SMILES C\C(=C/C=C/C(=C/C(=O)O)/C)\C=C\C1=C(C)C(O)CCC1(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H28O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13,18,21H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13+
Formula Weight 316.2038
Chemical Name or Material all-trans 4-Hydroxy Retinoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.