CAS RN 66592-72-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac all-trans 4-Hydroxy Retinoic Acid, TRC
CAS: 66592-72-1 Molecular Formula: C20 H28 O3 Molecular Weight (g/mol): 316.43 Synonym: Retinoic acid, 4-hydroxy-,4-Hydroxyretinoic acid,4-Hydroxy-all-trans-retinoic acid,4-OH-atRA,all-trans-4-Hydroxyretinoic acid IUPAC Name: (2E,4E,6E,8E)-9-(3-hydroxy-2,6,6-trimethylcyclohexen-1-yl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid SMILES: C\C(=C/C=C/C(=C/C(=O)O)/C)\C=C\C1=C(C)C(O)CCC1(C)C