missing translation for 'onlineSavingsMsg'
Learn More

rac-3-Hydroxyoctadecanoic Acid, TRC

Product Code. 30407464
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407464 100 mg 100mg
30395902 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407464 Supplier Toronto Research Chemicals Supplier No. TRCH943060100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17773-30-7
Molecular Formula C18H36O3
Molecular Weight (g/mol) 300.48
Synonym rac-3-Hydroxystearic Acid, rac-β-Hydroxyoctadecanoic Acid, rac-β-Hydroxystearic Acid
SMILES OC(CC(O)=O)CC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-17(19)16-18(20)21/h17,19H,2-16H2,1H3,(H,20,21)
Chemical Name or Material rac-3-Hydroxyoctadecanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.