CAS RN 17773-30-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac-3-Hydroxyoctadecanoic Acid, TRC
CAS: 17773-30-7 Molecular Formula: C18H36O3 Molecular Weight (g/mol): 300.48 Synonym: rac-3-Hydroxystearic Acid,rac-β-Hydroxyoctadecanoic Acid,rac-β-Hydroxystearic Acid SMILES: OC(CC(O)=O)CC