Learn More
Rabies Virus Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.
£95.06 - £530.40
Specifications
| Antigen | Rabies Virus Ab-1 |
|---|---|
| Clone | Rab-50 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Concentrated |
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|
12611617
|
Epredia
MS-549-P0 |
100 μL | 200μg/mL | ||||||
|
12621617
|
Epredia
MS-549-P1 |
500 μL | 200μg/mL | ||||||
|
12601617
|
Epredia
MS-549-P |
1 mL | 200μg/mL | ||||||
|
12631617
|
Epredia
MS-549-P1ABX |
100 μL | 1mg/mL | ||||||
|
12641617
|
Epredia
MS-549-PABX |
200 μL | 1mg/mL | ||||||
Description
Host Species: Mouse
Clone: Rab-50
Isotype: IgG2b/κ
Species Reactivity: Rabies virus of strains SAD-Vnukovo and Pitman Moore
Epitope: Not determined
Immunogen: Rabies Virus of strains SAD-Vnukovo and Pitman Moore Rab-50 antibody reacts with viral glycoprotein of rabies virus strains SAD-Vnukovo and Pitman-Moore. It can be used for detecting rabies virus by ELISA and Western. It is capable of neutralizing rabies virus.
Recommended for:
- ELISA
- Neutralization in vitro
Note:
USA: RUO; Int'l: RUO
Specifications
| Rabies Virus Ab-1 | |
| Monoclonal | |
| Concentrated | |
| RUO | |
| Rabies virus | |
| IgG2b κ |
| Rab-50 | |
| Unconjugated | |
| Mouse | |
| Infectious Disease | |
| Rabies Virus of strains SAD-Vnukovo and Pitman Moore | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.