Learn More
Rabies Virus Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-549-P1ABX
Description
Host Species: Mouse
Clone: Rab-50
Isotype: IgG2b/κ
Species Reactivity: Rabies virus of strains SAD-Vnukovo and Pitman Moore
Epitope: Not determined
Immunogen: Rabies Virus of strains SAD-Vnukovo and Pitman Moore Rab-50 antibody reacts with viral glycoprotein of rabies virus strains SAD-Vnukovo and Pitman-Moore. It can be used for detecting rabies virus by ELISA and Western. It is capable of neutralizing rabies virus.
Recommended for:
- ELISA
- Neutralization in vitro
Note:
USA: RUO; Int'l: RUO
Specifications
| Rabies Virus Ab-1 | |
| Rab-50 | |
| Unconjugated | |
| Rabies Virus of strains SAD-Vnukovo and Pitman Moore | |
| RUO | |
| Primary | |
| Concentrated |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Infectious Disease | |
| Rabies virus | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.