missing translation for 'onlineSavingsMsg'
Learn More

RABEPK/p40 Mouse anti-Human, Mouse, Rat, Clone: 1E11A3, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen RABEPK/p40
Clone 1E11A3
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808675
View Documents
Proteintech
66622-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16898665
View Documents
Proteintech
66622-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
Specifications

Specifications

RABEPK/p40
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
40 kDa Rab9 effector protein, p40, RAB9P40, RABEPK
Rabepk
IgG1
Protein G
Antibody
1E11A3
Western Blot
Unconjugated
Mouse
Mouse, Human, Rat
Q4V8F4, Q7Z6M1, Q8VCH5
10244, 227746, 296649
RABEPK/p40 Fusion Protein Ag7796
Primary
-20°C
RABEPK
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.