missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABEPK/p40 Mouse anti-Human, Mouse, Rat, Clone: 1E11A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66622-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.Specifications
| RABEPK/p40 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4V8F4, Q7Z6M1, Q8VCH5 | |
| Rabepk | |
| RABEPK/p40 Fusion Protein Ag7796 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1E11A3 | |
| Unconjugated | |
| RABEPK | |
| 40 kDa Rab9 effector protein, p40, RAB9P40, RABEPK | |
| Mouse | |
| Protein G | |
| RUO | |
| 10244, 227746, 296649 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction