missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit TNF-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £346.00
Specifications
| Antigen | TNF-alpha |
|---|---|
| Clone | 610325 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781929
![]() |
R&D Systems
MAB56701-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18169340
|
R&D Systems
MAB56701 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Rabbit samples. It is validated for Western Blot.Specifications
| TNF-alpha | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rabbit | |
| P04924 | |
| 7124 | |
| E. coli-derived recombinant rabbit TNF-alpha Leu79-Leu234 Accession # P04924 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects rabbit TNF-alpha in direct ELISAs. In direct ELISAs, 50-100% cross-reactivity with recombinant cotton rat, feline, guinea pig, human, mouse, rat TNF-alpha , less than 10% cross-reactivity with recombinant canine, equine, rhesus TNF-alpha , and no cross-reactivity with recombinant bovine or porcine TNF-alpha is observed. No cross-reactivity with recombinant human (rh) 4-1BB Ligand, APRIL, rhBAFF, rhEDA, rhEDA-A2, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhLymphotoxin alpha 1 beta 2, rhLymphotoxin alpha 2 beta 1, rhOX40 Ligand, rhTRAIL, rhTRANCE, rhTWEAK, rhVEGI, recombinant mouse (rm) 4-1BB Ligand, rmBAFF, rmFas Ligand, rmGITR Ligand, rmLIGHT, rmOX40 Ligand, rmTRAIL, rmTRAIL-2 Ligand, rmTRANCE, or rmTWEAK is observed. |
| 610325 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| TNF | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title