missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit TNF-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB56701-SP
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Rabbit samples. It is validated for Western Blot.
Specifications
| TNF-alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P04924 | |
| TNF | |
| E. coli-derived recombinant rabbit TNF-alpha Leu79-Leu234 Accession # P04924 | |
| 25 μg | |
| Primary | |
| Detects rabbit TNF-alpha in direct ELISAs. In direct ELISAs, 50-100% cross-reactivity with recombinant cotton rat, feline, guinea pig, human, mouse, rat TNF-alpha , less than 10% cross-reactivity with recombinant canine, equine, rhesus TNF-alpha , and no cross-reactivity with recombinant bovine or porcine TNF-alpha is observed. No cross-reactivity with recombinant human (rh) 4-1BB Ligand, APRIL, rhBAFF, rhEDA, rhEDA-A2, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhLymphotoxin alpha 1 beta 2, rhLymphotoxin alpha 2 beta 1, rhOX40 Ligand, rhTRAIL, rhTRANCE, rhTWEAK, rhVEGI, recombinant mouse (rm) 4-1BB Ligand, rmBAFF, rmFas Ligand, rmGITR Ligand, rmLIGHT, rmOX40 Ligand, rmTRAIL, rmTRAIL-2 Ligand, rmTRANCE, or rmTWEAK is observed. | |
| Rabbit | |
| Purified |
| Western Blot | |
| 610325 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7124 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction