missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab5a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 12 publications
£176.00
Specifications
| Antigen | Rab5a |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry Free-Floating |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Rab5a Polyclonal specifically detects Rab5a in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.Specifications
| Rab5a | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry Free-Floating | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Q6FI44 | |
| 5868 | |
| Human Rab5 synthetic peptide conjugated to KLH; identical to dog Rab5 sequence over the residues | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endosome Markers, Signal Transduction | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A | |
| RAB5A | |
| IgG | |
| Protein A purified | |
| Detects approx 26kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title