missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab5a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NB120-13253-0.025mg
This item is not returnable.
View return policy
Description
Rab5a Polyclonal specifically detects Rab5a in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.
Specifications
| Rab5a | |
| Polyclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| RAB5A, member RAS oncogene family, RAB5RAS-associated protein RAB5A, ras-related protein Rab-5A | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Detects approx 26kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry Free-Floating | |
| Q6FI44 | |
| RAB5A | |
| Human Rab5 synthetic peptide conjugated to KLH; identical to dog Rab5 sequence over the residues | |
| 0.025 mg | |
| Endosome Markers, Signal Transduction | |
| 5868 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction