missing translation for 'onlineSavingsMsg'
Learn More

RAB4A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen RAB4A
Concentration 200 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16826683
View Documents
Proteintech
10347-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16816683
View Documents
Proteintech
10347-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RAB4A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAB4A. This antibody recognizes human, mouse, rat antigen. The RAB4A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.

Specifications

Specifications

RAB4A
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Human, Rat, Mouse
IgG
200 μg/mL
Polyclonal
Rabbit
5867
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.