Learn More
RAB4A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10347-1-AP-150UL
 View more versions of this product
Product Code. 16816683
  • £349.00 / 150µL
Description
The RAB4A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAB4A. This antibody recognizes human, mouse, rat antigen. The RAB4A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
RAB4A
Polyclonal
Unconjugated
150 μL
5867
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
200 μg/mL
Rabbit
Primary
Human, Rat, Mouse