missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB35 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RAB35 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808333
|
Proteintech
11329-2-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898323
|
Proteintech
11329-2-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB35 possesses GTPase activity.Specifications
| RAB35 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q15286, Q5U316, Q6PHN9 | |
| 11021, 288700, 77407 | |
| RAB35 Fusion Protein Ag1872 | |
| Primary | |
| -20°C | |
| RAB35 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GTP binding protein RAY, H ray, RAB1C, RAB35, Ras related protein Rab 1C, Ras related protein Rab 35, RAY | |
| Rab35 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title